Learn More
Invivogen HEK-Blue™ TSLP Cells, Monitors TSLP Induced STAT5 Activation, Colorimetric Bioassay, 1 vial
SDP
Catalog No. NC3444964
Encompass
The InvivoGen HEK-Blue™ TSLP Cells are a robust bioassay tool designed to analyze human TSLP signaling through STAT5 activation, utilizing a colorimetric detection method. Derived from HEK 293 cells, these cells offer high sensitivity and specificity for TSLP research applications, making them ideal for studying immune responses and inflammation. Their stable nature and pre-validation for cytokine testing differentiate them from other options, ensuring reliable and consistent results in research.
Catalog No. NC3444964 Supplier Invivogen Supplier No. HKBTSLP
Add to Cart
Add to Cart
The InvivoGen HEK-Blue™ TSLP Cells are a robust bioassay tool designed to analyze human TSLP signaling through STAT5 activation, utilizing a colorimetric detection method. Derived from HEK 293 cells, these cells offer high sensitivity and specificity for TSLP research applications, making them ideal for studying immune responses and inflammation. Their stable nature and pre-validation for cytokine testing differentiate them from other options, ensuring reliable and consistent results in research.