Learn More
Eurofins Discoverx PathHunter HEK 293 CXCR2 (IL8RB) β-Arrestin Cell Line, 2 vials. (93-0202C1)
Catalog No. 501999936
Encompass
The PathHunter CXCR2 (IL8RB) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CXCL8/IL8) based activation of CXCR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CXCR2 and EA-tagged β-Arrestin-2. Activation of CXCR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-199-9936 Supplier Eurofins Discoverx Supplier No. 930202C1
Add to Cart
Add to Cart
Request a Quote
The PathHunter CXCR2 (IL8RB) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CXCL8/IL8) based activation of CXCR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CXCR2 and EA-tagged β-Arrestin-2. Activation of CXCR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.