Learn More
Eurofins Discoverx PathHunter HEK 293 IL11R/IL6ST Dimerization Cell Line, 2 vials. (93-1028C1)
Catalog No. 502001879
Encompass
The PathHunter IL11R/IL6ST Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-11) induced dimerization of IL11R/IL6ST receptors. This product uses EFC technology. The cell line included overexpresses IL6ST target protein fused to PK alongside IL11R target protein fused to EA. Upon ligand-induced dimerization of IL11R/IL6ST, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-1879 Supplier Eurofins Discoverx Supplier No. 931028C1
Add to Cart
Add to Cart
Request a Quote
The PathHunter IL11R/IL6ST Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-11) induced dimerization of IL11R/IL6ST receptors. This product uses EFC technology. The cell line included overexpresses IL6ST target protein fused to PK alongside IL11R target protein fused to EA. Upon ligand-induced dimerization of IL11R/IL6ST, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.