Learn More
Eurofins Discoverx PathHunter HEK 293 IL17RA/IL17RC Dimerization Cell Line, 2 vials. (93-0931C1)
Catalog No. 502001592
Encompass
The PathHunter IL17RA/IL17RC Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-17a) induced dimerization of IL17RA/IL17RC receptors. This product uses EFC technology. The cell line included overexpresses IL17RC target protein fused to PK alongside IL17RA target protein fused to EA. Upon ligand-induced dimerization of IL17RA/IL17RC, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-1592 Supplier Eurofins Discoverx Supplier No. 930931C1
Add to Cart
Add to Cart
Request a Quote
The PathHunter IL17RA/IL17RC Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-17a) induced dimerization of IL17RA/IL17RC receptors. This product uses EFC technology. The cell line included overexpresses IL17RC target protein fused to PK alongside IL17RA target protein fused to EA. Upon ligand-induced dimerization of IL17RA/IL17RC, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.