Learn More
Eurofins Discoverx PathHunter HEK 293 IL3RA/CD131 Dimerization Cell Line, 2 vials. (93-0969C1)
Supplier:  Eurofins Discoverx 930969C1
Encompass
The PathHunter IL3RA/CD131 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-3) induced dimerization of IL3RA/CD131 receptors. This product uses EFC technology. The cell line included overexpresses IL3RA target protein fused to PK alongside CD131 target protein fused to EA. Upon ligand-induced dimerization of IL3RA/CD131, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-1712
Add to Cart
Request a Quote