Learn More
Eurofins Discoverx InCELL Hunter HEK 293 TAF1L(2) Bromodomain Cell Line, 2 vials. (96-0058C1)
Catalog No. 502004455
Encompass
The InCELL Hunter HEK 293 TAF1L(2) Bromodomain Cell Line is a stable, engineered cell line for use in studying small molecule drug candidates that engage with regulators of epigenetics pathways. This cell line enables assessment of compound-engagement-based stabilization of TAF1L(2) via detection of TAF1L(2) levels. This product uses EFC technology. The cell line included overexpresses TAF1L(2) target protein fused to ProLabel. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/epigenetics.
Catalog No. 50-200-4455 Supplier Eurofins Discoverx Supplier No. 960058C1
Add to Cart
Add to Cart
Request a Quote
The InCELL Hunter HEK 293 TAF1L(2) Bromodomain Cell Line is a stable, engineered cell line for use in studying small molecule drug candidates that engage with regulators of epigenetics pathways. This cell line enables assessment of compound-engagement-based stabilization of TAF1L(2) via detection of TAF1L(2) levels. This product uses EFC technology. The cell line included overexpresses TAF1L(2) target protein fused to ProLabel. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/epigenetics.