Learn More
Eurofins Discoverx PathHunter HEK 293 TPOR/TPOR Dimerization Cell Line, 2 vials. (93-1108C1)
Catalog No. 502002127
Encompass
The PathHunter TPOR/TPOR Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. TPO) induced dimerization of TPOR/TPOR receptors. This product uses EFC technology. The cell line included overexpresses TPOR target protein fused to PK alongside TPOR target protein fused to EA. Upon ligand-induced dimerization of TPOR/TPOR, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-2127 Supplier Eurofins Discoverx Supplier No. 931108C1
Add to Cart
Add to Cart
Request a Quote
The PathHunter TPOR/TPOR Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. TPO) induced dimerization of TPOR/TPOR receptors. This product uses EFC technology. The cell line included overexpresses TPOR target protein fused to PK alongside TPOR target protein fused to EA. Upon ligand-induced dimerization of TPOR/TPOR, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.