Learn More
Genscript Corporation HEK293/5-HT2C Stable Cell Line
Catalog No. 5000058383
Encompass
5-Hydroxytryptamine (5-HT also commonly known as serotonin) is synthesized in enterochromaffin cells in the intestine and in serotonergic nerve terminals In the periphery 5-HT mediates gastrointestinal motility platelet aggregation and contraction of blood vessels Many functions of the central nervous system are influenced by 5-HT including sleep motor activity sensory perception arousal and appetite A family of 12 GPCRs and one ion channel mediate the biological effects of 5-HT (Hoyer et al 1994) 5HT2C is expressed in the brain and spinal cord especially the choroid plexus 5HT2C receptor agonists may have important clinical value in the treatment of mental and eating disorders such as depression panic anxiety OCD bulimia and obesity GenScript s cloned human 5-HT2C expressing cell line is generated in the CHO-K1 host
Catalog No. 50-000-58383 Supplier Genscript Corporation Supplier No. M00319
Add to Cart
Add to Cart
Request a Quote
5-Hydroxytryptamine (5-HT also commonly known as serotonin) is synthesized in enterochromaffin cells in the intestine and in serotonergic nerve terminals In the periphery 5-HT mediates gastrointestinal motility platelet aggregation and contraction of blood vessels Many functions of the central nervous system are influenced by 5-HT including sleep motor activity sensory perception arousal and appetite A family of 12 GPCRs and one ion channel mediate the biological effects of 5-HT (Hoyer et al 1994) 5HT2C is expressed in the brain and spinal cord especially the choroid plexus 5HT2C receptor agonists may have important clinical value in the treatment of mental and eating disorders such as depression panic anxiety OCD bulimia and obesity GenScript s cloned human 5-HT2C expressing cell line is generated in the CHO-K1 host