Learn More
Genscript Corporation HEK293/AT1 Stable Cell Line
Catalog No. 5000057445
Encompass
Recombinant HEK293 cells stably overexpress human angiotensin II receptor type 1 (AT1) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57445 Supplier Genscript Corporation Supplier No. M00458
Add to Cart
Add to Cart
Request a Quote
Recombinant HEK293 cells stably overexpress human angiotensin II receptor type 1 (AT1) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways