Learn More
Eurofins Discoverx InCELL Hunter HEK 293 BAZ2A Bromodomain Cell Line, 2 vials. (96-0074C1)
Supplier:  Eurofins Discoverx 960074C1
Encompass
The InCELL Hunter HEK 293 BAZ2A Bromodomain Cell Line is a stable, engineered cell line for use in studying small molecule drug candidates that engage with regulators of epigenetics pathways. This cell line enables assessment of compound-engagement-based stabilization of BAZ2A via detection of BAZ2A levels. This product uses EFC technology. The cell line included overexpresses BAZ2A target protein fused to ProLabel. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/epigenetics.
Catalog No. 50-200-4515
Add to Cart
Request a Quote