Learn More
Invivogen HEK-Blue™ IFN-γ Cells, Detects Bioactive IFN-γ, JAK/STAT1 Pathway Activation, 1 vial
SDP
Catalog No. NC1906394
Encompass
InvivoGen HKB-IFNG HEK-Blue™ IFN-γ Cells are engineered to detect human interferon-gamma (IFN-γ) via the JAK/STAT1 signaling pathway. These specialized HEK293 reporter cells enable quick screening of activatory or inhibitory molecules impacting STAT1 stimulation. Stably transfected with the STAT1 gene, they ensure reliability in bioassays, distinguishing them from standard cell lines. This system aids in studying immune responses and therapeutic interventions.
Catalog No. NC1906394 Supplier Invivogen Supplier No. HKBIFNG
InvivoGen HKB-IFNG HEK-Blue™ IFN-γ Cells are engineered to detect human interferon-gamma (IFN-γ) via the JAK/STAT1 signaling pathway. These specialized HEK293 reporter cells enable quick screening of activatory or inhibitory molecules impacting STAT1 stimulation. Stably transfected with the STAT1 gene, they ensure reliability in bioassays, distinguishing them from standard cell lines. This system aids in studying immune responses and therapeutic interventions.