Learn More
Genscript Corporation HEK293/CLDN18.2 Stable Cell Li
Catalog No. 5000056783
Encompass
Claudins are a family of proteins which play a role in maintaining tight cell junctions Differentclaudin subtypes are expressed on different tissues Claudin18 isoform 2 (CLDN18 2) expressionin normal tissues is restricted to cells of the gastric mucosa and is absent from other healthytissues However CLDN 18 2 is expressed in 70% of primary gastric adenocarcinomas and itsmetastases (101) It has also beenshown to be expressed in a number of other cancers including pancreatic (50%) esophagus (30%) and NSCLC (25%)The restricted expressionmakes it a potential drug target for the treatment of gastric and pancreatic adenocarcinoma asevidenced by effortsto target CLDN18 2 via naked antibody and CAR-T modalities
Catalog No. 50-000-56783 Supplier Genscript Corporation Supplier No. M00917
Add to Cart
Add to Cart
Request a Quote
Claudins are a family of proteins which play a role in maintaining tight cell junctions Differentclaudin subtypes are expressed on different tissues Claudin18 isoform 2 (CLDN18 2) expressionin normal tissues is restricted to cells of the gastric mucosa and is absent from other healthytissues However CLDN 18 2 is expressed in 70% of primary gastric adenocarcinomas and itsmetastases (101) It has also beenshown to be expressed in a number of other cancers including pancreatic (50%) esophagus (30%) and NSCLC (25%)The restricted expressionmakes it a potential drug target for the treatment of gastric and pancreatic adenocarcinoma asevidenced by effortsto target CLDN18 2 via naked antibody and CAR-T modalities