Learn More
Eurofins Discoverx InCELL Hunter HEK 293 FGR Kinase Binding Cell Line, 2 vials. (96-0043C1)
Catalog No. 502004293
Encompass
The InCELL Hunter HEK 293 FGR Kinase Binding Cell Line is a stable, engineered cell line for use in studying small molecule drug candidates that engage with kinases. This cell line enables assessment of compound-engagement-based stabilization of FGR via detection of FGR levels. This product uses EFC technology. The cell line included overexpresses FGR target protein fused to ProLabel. In the absence of compound that binds and stabilizes the target, this protein is denatured by the heat treatment and no EFC signal is produced. When compound engages and heat-stabilizes the target, EA contained in the detection reagent produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-4293 Supplier Eurofins Discoverx Supplier No. 960043C1
Add to Cart
Add to Cart
Request a Quote
The InCELL Hunter HEK 293 FGR Kinase Binding Cell Line is a stable, engineered cell line for use in studying small molecule drug candidates that engage with kinases. This cell line enables assessment of compound-engagement-based stabilization of FGR via detection of FGR levels. This product uses EFC technology. The cell line included overexpresses FGR target protein fused to ProLabel. In the absence of compound that binds and stabilizes the target, this protein is denatured by the heat treatment and no EFC signal is produced. When compound engages and heat-stabilizes the target, EA contained in the detection reagent produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.