Learn More
Genscript Corporation HEK293/FP/G15 Stable Cell Lin
Catalog No. 5000057079
Encompass
Prostaglandin F (2-alpha) is known as a potent luteolytic agent It is involved in modulating intraocular pressure and smooth muscle contraction in uterus Its effects on cells are mediated through specific interaction with the Prostanoid receptor FP which is a 359-amino acid protein Having 7 putative transmembrane domains FP resembles the characteristic of G protein coupled receptors Knockout studies in mice suggest that the interaction of PGF2-alpha with this receptor may initiate parturition in ovarian luteal cells and thereupon induce luteolysis
Catalog No. 50-000-57079 Supplier Genscript Corporation Supplier No. M00155
Add to Cart
Add to Cart
Request a Quote
Prostaglandin F (2-alpha) is known as a potent luteolytic agent It is involved in modulating intraocular pressure and smooth muscle contraction in uterus Its effects on cells are mediated through specific interaction with the Prostanoid receptor FP which is a 359-amino acid protein Having 7 putative transmembrane domains FP resembles the characteristic of G protein coupled receptors Knockout studies in mice suggest that the interaction of PGF2-alpha with this receptor may initiate parturition in ovarian luteal cells and thereupon induce luteolysis