Learn More
Genscript Corporation HEK293/GCGR/G15 Stable Cell L
Catalog No. 5000058733
Encompass
Recombinant HEK293 cells stably overexpress human glucagon receptor (GCGR) on the surface and contain high levels of G protein G s to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-58733 Supplier Genscript Corporation Supplier No. M00422
Add to Cart
Add to Cart
Request a Quote
Recombinant HEK293 cells stably overexpress human glucagon receptor (GCGR) on the surface and contain high levels of G protein G s to couple with the receptor in downstream signaling pathways