Learn More
ACROBiosystems HEK293 Human GIPR Stable Cell Line, High Expression, 2 Vials
Catalog No. NC3761288
Encompass_Preferred
HEK293/Human GIPR stable cell line engineered to express full-length human GIPR at high levels for robust cell-based assays and agonist screening. Surface expression is validated by flow cytometry. Supplied frozen with QC data and licensing support for commercial use.
  • Supplied as frozen vials for long-term storage in liquid nitrogen.
  • Each vial contains at least 5 x 10^6 cells in 1 mL serum-free cryopreservation medium.
  • High receptor expression for a larger assay window and reproducible results.
  • Validated for cell-based GIPR binding assays and cAMP accumulation agonist screening.
  • Includes quality control data and documented passage stability.
Catalog No. NC3761288 Supplier ACROBiosystems Supplier No. CHEKATP2062VIALS
HEK293/Human GIPR stable cell line engineered to express full-length human GIPR at high levels for robust cell-based assays and agonist screening. Surface expression is validated by flow cytometry. Supplied frozen with QC data and licensing support for commercial use.
  • Supplied as frozen vials for long-term storage in liquid nitrogen.
  • Each vial contains at least 5 x 10^6 cells in 1 mL serum-free cryopreservation medium.
  • High receptor expression for a larger assay window and reproducible results.
  • Validated for cell-based GIPR binding assays and cAMP accumulation agonist screening.
  • Includes quality control data and documented passage stability.