Learn More
ACROBiosystems HEK293/Human GIPR Stable Cell Line (Low Expression), cryopreserved, 2 vials
Catalog No. NC3761290
Encompass_Preferred
HEK293/Human GIPR Stable Cell Line (Low Expression) is an adherent HEK293-derived cell line engineered to express full-length human GIPR at low levels. Supplied frozen in two vials, the cell line is intended for cell-based GIPR binding assays and functional screening of GIPR agonists using cAMP accumulation assays.
  • HEK293-derived adherent cell line expressing full-length human GIPR at low levels.
  • Supplied as two cryopreserved vials, each containing ≥5 x 10^6 cells in 1 mL serum-free cryopreservation medium.
  • Selection marker: hygromycin B (20 μg/mL).
  • Complete growth medium: DMEM with 10% fetal bovine serum.
  • Mycoplasma and sterility tested negative.
  • Stored frozen in liquid nitrogen for long-term preservation.
  • Suitable for cell-based GIPR binding and cAMP accumulation assays for agonist screening.
Catalog No. NC3761290 Supplier ACROBiosystems Supplier No. CHEKATP2082VIALS
HEK293/Human GIPR Stable Cell Line (Low Expression) is an adherent HEK293-derived cell line engineered to express full-length human GIPR at low levels. Supplied frozen in two vials, the cell line is intended for cell-based GIPR binding assays and functional screening of GIPR agonists using cAMP accumulation assays.
  • HEK293-derived adherent cell line expressing full-length human GIPR at low levels.
  • Supplied as two cryopreserved vials, each containing ≥5 x 10^6 cells in 1 mL serum-free cryopreservation medium.
  • Selection marker: hygromycin B (20 μg/mL).
  • Complete growth medium: DMEM with 10% fetal bovine serum.
  • Mycoplasma and sterility tested negative.
  • Stored frozen in liquid nitrogen for long-term preservation.
  • Suitable for cell-based GIPR binding and cAMP accumulation assays for agonist screening.