Learn More
ACROBiosystems HEK293 Human GIPR Stable Cell Line (Medium Expression), 2 vials (1 mL, ≥5x10^6 cells/vial)
Catalog No. NC3761283
Encompass_Preferred
HEK293 human GIPR stable cell line supplied as two cryopreserved vials. Each vial contains at least 5x10^6 cells in 1 mL serum-free cryopreservation medium and is stored in liquid nitrogen. Surface expression of full-length human GIPR has been validated by flow cytometry. The cell line is intended for cell-based binding and functional assays, including cAMP accumulation assays.
  • Engineered to express full-length human GIPR.
  • Medium expression level suitable for receptor pharmacology and screening.
  • Delivered as two cryopreserved vials, 1 mL each.
  • Contains at least 5x10^6 viable cells per vial.
  • Frozen in liquid nitrogen for long-term storage.
  • Validated by flow cytometry for surface receptor expression.
  • Suitable for cell-based binding and cAMP accumulation functional assays.
Catalog No. NC3761283 Supplier ACROBiosystems Supplier No. CHEKATP2072VIALS
HEK293 human GIPR stable cell line supplied as two cryopreserved vials. Each vial contains at least 5x10^6 cells in 1 mL serum-free cryopreservation medium and is stored in liquid nitrogen. Surface expression of full-length human GIPR has been validated by flow cytometry. The cell line is intended for cell-based binding and functional assays, including cAMP accumulation assays.
  • Engineered to express full-length human GIPR.
  • Medium expression level suitable for receptor pharmacology and screening.
  • Delivered as two cryopreserved vials, 1 mL each.
  • Contains at least 5x10^6 viable cells per vial.
  • Frozen in liquid nitrogen for long-term storage.
  • Validated by flow cytometry for surface receptor expression.
  • Suitable for cell-based binding and cAMP accumulation functional assays.