Learn More
Eurofins Discoverx PathHunter HEK 293 IFNAR1/IFNAR2 Dimerization Cell Line, 2 vials. (93-1097C1)
Catalog No. 502002050
Encompass
The PathHunter IFNAR1/IFNAR2 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IFN-beta) induced dimerization of IFNAR1/IFNAR2 receptors. This product uses EFC technology. The cell line included overexpresses IFNAR2 target protein fused to PK alongside IFNAR1 target protein fused to EA. Upon ligand-induced dimerization of IFNAR1/IFNAR2, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-2050 Supplier Eurofins Discoverx Supplier No. 931097C1
Add to Cart
Add to Cart
Request a Quote
The PathHunter IFNAR1/IFNAR2 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IFN-beta) induced dimerization of IFNAR1/IFNAR2 receptors. This product uses EFC technology. The cell line included overexpresses IFNAR2 target protein fused to PK alongside IFNAR1 target protein fused to EA. Upon ligand-induced dimerization of IFNAR1/IFNAR2, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.