Learn More
Eurofins Discoverx PathHunter HEK 293 IL9R/IL2RG Dimerization Cell Line, 2 vials. (93-1036C1)
Catalog No. 502001900
Encompass
The PathHunter IL9R/IL2RG Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-9) induced dimerization of IL9R/IL2RG receptors. This product uses EFC technology. The cell line included overexpresses IL2RG target protein fused to PK alongside IL9R target protein fused to EA. Upon ligand-induced dimerization of IL9R/IL2RG, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-1900 Supplier Eurofins Discoverx Supplier No. 931036C1
Add to Cart
Add to Cart
Request a Quote
The PathHunter IL9R/IL2RG Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-9) induced dimerization of IL9R/IL2RG receptors. This product uses EFC technology. The cell line included overexpresses IL2RG target protein fused to PK alongside IL9R target protein fused to EA. Upon ligand-induced dimerization of IL9R/IL2RG, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.