Learn More
Gentarget HEK293-mCherry Stable Cell Line, 2 x 10^6 cells/vial, Red Fluorescent Protein, Enhanced CMV Promoter, Puromycin Resistance, Low Cytotoxicity
Catalog No. NC3237248
Encompass
HEK293-mCherry Stable Cell Line, 2 x 10^6 cells/vial, Red Fluorescent Protein, Enhanced CMV Promoter, Puromycin Resistance, Low Cytotoxicity
Catalog No. NC3237248 Supplier Gentarget Supplier No. SC097
HEK293-mCherry Stable Cell Line, 2 x 10^6 cells/vial, Red Fluorescent Protein, Enhanced CMV Promoter, Puromycin Resistance, Low Cytotoxicity