Learn More
ACROBiosystems HEK293 Membrane-Bound human TL1A Stable Cell Line, 2 vials, 1 mL each, at least 5x10^6 cells per vial, adherent, Puromycin selection, DMEM + 10% FBS
Catalog No. NC3727275
Encompass_Preferred
The HEK293/Membrane-Bound human TL1A Stable Cell Line is a genetically engineered cell line optimized for robust TL1A binding assays. This adherent cell line expresses membrane-bound human TL1A after specific amino acid deletion. With comprehensive application data and stringent quality control, this cell line facilitates reliable bioassays supported by validated stability data.
  • Cell line optimally designed for TL1A binding assays
  • Membrane-bound human TL1A expressed
  • Flow cytometry used for expression confirmation
  • Adherent growth properties
  • Selection marker: Puromycin, 2 μg/mL
  • Complete growth medium: DMEM + 10% FBS
  • Mycoplasma and sterility tested: negative results
  • Comprehensive quality control records available
Catalog No. NC3727275 Supplier ACROBiosystems Supplier No. CHEKATP1982VIALS
The HEK293/Membrane-Bound human TL1A Stable Cell Line is a genetically engineered cell line optimized for robust TL1A binding assays. This adherent cell line expresses membrane-bound human TL1A after specific amino acid deletion. With comprehensive application data and stringent quality control, this cell line facilitates reliable bioassays supported by validated stability data.
  • Cell line optimally designed for TL1A binding assays
  • Membrane-bound human TL1A expressed
  • Flow cytometry used for expression confirmation
  • Adherent growth properties
  • Selection marker: Puromycin, 2 μg/mL
  • Complete growth medium: DMEM + 10% FBS
  • Mycoplasma and sterility tested: negative results
  • Comprehensive quality control records available