Learn More
Genscript Corporation HEK293/PD-L1 Stable Cell Line
Catalog No. 5000057652
Encompass
Programmed death-ligand 1 (PD-L1) also known as cluster of differentiation 274 (CD274) or B7 homolog 1 (B7-H1) is a protein that in humans is encoded by the CD274 gene The formation of the PD-1 receptor / PD-L1 or B7 1 receptor /PD-L1 ligand complex transmits an inhibitory signal which reduces the proliferation of these CD8 T cells at the lymph nodes PD-1 is also able to control the accumulation of foreign antigen specific T cells in the lymph nodes through apoptosis which is further mediated by a lower regulation of the gene Bcl-2
Catalog No. 50-000-57652 Supplier Genscript Corporation Supplier No. M00544
Add to Cart
Add to Cart
Request a Quote
Programmed death-ligand 1 (PD-L1) also known as cluster of differentiation 274 (CD274) or B7 homolog 1 (B7-H1) is a protein that in humans is encoded by the CD274 gene The formation of the PD-1 receptor / PD-L1 or B7 1 receptor /PD-L1 ligand complex transmits an inhibitory signal which reduces the proliferation of these CD8 T cells at the lymph nodes PD-1 is also able to control the accumulation of foreign antigen specific T cells in the lymph nodes through apoptosis which is further mediated by a lower regulation of the gene Bcl-2