Learn More
Eurofins Discoverx PathHunter HEK 293 RELA Nuclear Translocation Cell Line, 2 vials. (93-0907C1)
Catalog No. 502001541
Encompass
The PathHunter HEK 293 RELA Nuclear Translocation Cell Line is a stable, engineered cell line for use in studying drug candidates affecting the NFκB pathway. This cell line enables assessment of ligand (e.g. IL-1α) based pathway activation via detection of resulting RELA nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLink-tagged RELA and nuclear localized EA. Upon pathway activation, induced nuclear translocation of RELA leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/pathways.
Catalog No. 50-200-1541 Supplier Eurofins Discoverx Supplier No. 930907C1
Add to Cart
Add to Cart
Request a Quote
The PathHunter HEK 293 RELA Nuclear Translocation Cell Line is a stable, engineered cell line for use in studying drug candidates affecting the NFκB pathway. This cell line enables assessment of ligand (e.g. IL-1α) based pathway activation via detection of resulting RELA nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLink-tagged RELA and nuclear localized EA. Upon pathway activation, induced nuclear translocation of RELA leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/pathways.