Learn More
Invivogen InvivoGen HEK-Dual™ Cells, Monitors NF-κB and IRF Pathways, Human Mycoplasma-free, 4 units
SDP
Supplier:  Invivogen HKDNFIS
Encompass
InvivoGen HEK-Dual™ Reporter Cells (NF-κB / IRF) are engineered HEK293-derived cells that enable simultaneous monitoring of NF-κB and IRF signaling pathways. This dual-reporter system provides a rapid and sensitive method for studying innate immunity and screening PRR modulators, distinguishing it from other products by its ability to non-destructively measure secreted reporter proteins.
Catalog No. NC2541708