Learn More
Gentarget HEK293 / uGFP (unstable GFP) Stable Cell Line
Catalog No. NC2880673
Encompass
This stable cell line is transformed from the adhesive enhanced HEK293 cells, stably expressing the unstable GFP (click to see sequence) fluorescent protein. It was established by transduction of the destabilized GFP expression lentivirus containing a Puromycin resistance marker. GFP is constitutively expressed at high-level under the enhanced CMV promoter (suCMV). The following expression construct was integrated into the cell line’s genome.
Catalog No. NC2880673 Supplier Gentarget Supplier No. SC058
Add to Cart
Add to Cart
This stable cell line is transformed from the adhesive enhanced HEK293 cells, stably expressing the unstable GFP (click to see sequence) fluorescent protein. It was established by transduction of the destabilized GFP expression lentivirus containing a Puromycin resistance marker. GFP is constitutively expressed at high-level under the enhanced CMV promoter (suCMV). The following expression construct was integrated into the cell line’s genome.