Learn More
ATCC HEK293S GnTI-; Embryonic Kidney; Human (Homo sapiens)
Catalog No. 502383284
Encompass
HEK293S GnTI? is a cell line exhibiting epithelial-like morphology that was isolated from the kidney of an embryo. It was established by methanesulfonate mutagenesis followed by ricin selection. This cell line has been successfully used to overexpress a wide variety of mammalian membrane proteins.
Catalog No. 50-238-3284 Supplier ATCC Supplier No. CRL3022
HEK293S GnTI? is a cell line exhibiting epithelial-like morphology that was isolated from the kidney of an embryo. It was established by methanesulfonate mutagenesis followed by ricin selection. This cell line has been successfully used to overexpress a wide variety of mammalian membrane proteins.