Learn More
CYTION HEK293T/17 Cells 305117
Catalog No. NC2952825
Encompass
The 293T/17 cell line is an immortalized variant of the HEK293 line, derived from human embryonic kidney cells and extensively utilized in research, particularly in the study and production of retroviral and lentiviral vectors. This cell line has been modified to express the SV40 large T antigen, enhancing its utility in viral vector production. The expression of the SV40 large T antigen is a key feature that allows these cells to replicate plasmids containing the SV40 origin of replication, significantly increasing the yield of plasmid DNA in transient transfection procedures. This feature is particularly beneficial for the production of viral vectors.
Catalog No. NC2952825 Supplier CYTION Supplier No. 305117
The 293T/17 cell line is an immortalized variant of the HEK293 line, derived from human embryonic kidney cells and extensively utilized in research, particularly in the study and production of retroviral and lentiviral vectors. This cell line has been modified to express the SV40 large T antigen, enhancing its utility in viral vector production. The expression of the SV40 large T antigen is a key feature that allows these cells to replicate plasmids containing the SV40 origin of replication, significantly increasing the yield of plasmid DNA in transient transfection procedures. This feature is particularly beneficial for the production of viral vectors.