Learn More
Genscript Corporation HEK293T/Spike Stable Cell Line
Catalog No. 5000059228
Encompass
Recombinant HEK293T cells stably overexpress coronavirus SARS-CoV-2 spike protein on the surface The surface expression of SARS-CoV-2 spike protein is validated by FACS analysis This stable cell line product is designed for screening antibodies against SARS-CoV-2 spike protein as well as for measuring binding affinity and stability of antibody based biologics that bind with spike protein GenScript also offers spike protein expressing CHO-K1 stable cell line (Cat No M00803) for SARS-CoV-2 study
Catalog No. 50-000-59228 Supplier Genscript Corporation Supplier No. M00804
Add to Cart
Add to Cart
Request a Quote
Recombinant HEK293T cells stably overexpress coronavirus SARS-CoV-2 spike protein on the surface The surface expression of SARS-CoV-2 spike protein is validated by FACS analysis This stable cell line product is designed for screening antibodies against SARS-CoV-2 spike protein as well as for measuring binding affinity and stability of antibody based biologics that bind with spike protein GenScript also offers spike protein expressing CHO-K1 stable cell line (Cat No M00803) for SARS-CoV-2 study