Learn More
MOLTOX Molecular Toxicology Inc Hektoen Enteric Agar, 20 mL/plate, 10 plates/sleeve, Store in Refrigerator/Dark
SDP
Catalog No. 50948808
Encompass_Preferred
Hektoen Enteric Agar is a fairly selective medium used for the isolation and cultivation of gram-negative enteric microorganisms, particularly Shigella. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-808 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S008
Hektoen Enteric Agar is a fairly selective medium used for the isolation and cultivation of gram-negative enteric microorganisms, particularly Shigella. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.