Learn More
NEOGEN Hektoen Enteric (HE) Agar, 10 kg
SDP
Catalog No. 502015173
Encompass_Preferred
Hektoen Enteric Agar is used for the isolation and differentiation of enteric pathogens. Hektoen Enteric Agar is not intended for use in the diagnosis of disease or other conditions in humans. Hektoen Enteric Agar was developed in 1967 by King and Metzger. Compared to other enteric differentiating media commonly used, Hektoen Enteric Agar increased the isolation rate of Salmonella spp. and Shigella spp. This was accomplished by increasing the carbohydrate and peptone content of the medium in order to counteract the inhibitory effects of bile salts and indicators. King and Metzger formulated a medium that slightly inhibited growth of Salmonella and Shigella, while inhibiting Gram-positive microorganisms. Enzymatic Digest of Animal Tissue provides nitrogen, carbon, and amino acids required for organism growth.For more information visit https://www.neogen.com/solutions/Microbiology/hektoen-enteric-agar
Catalog No. 50-201-5173 Supplier NEOGEN Supplier No. 700002973
Hektoen Enteric Agar is used for the isolation and differentiation of enteric pathogens. Hektoen Enteric Agar is not intended for use in the diagnosis of disease or other conditions in humans. Hektoen Enteric Agar was developed in 1967 by King and Metzger. Compared to other enteric differentiating media commonly used, Hektoen Enteric Agar increased the isolation rate of Salmonella spp. and Shigella spp. This was accomplished by increasing the carbohydrate and peptone content of the medium in order to counteract the inhibitory effects of bile salts and indicators. King and Metzger formulated a medium that slightly inhibited growth of Salmonella and Shigella, while inhibiting Gram-positive microorganisms. Enzymatic Digest of Animal Tissue provides nitrogen, carbon, and amino acids required for organism growth.For more information visit https://www.neogen.com/solutions/Microbiology/hektoen-enteric-agar