Learn More
ATCC HEL 92.1.7; Erythroleukemia; Human (Homo sapiens)
Supplier:  ATCC TIB180
Encompass
HEL 92.1.7 is a erythroblast cell line isolated from the bone marrow of a 30-year-old, White, male patient with erthroleukemia. This cell line can be used in immune system disorder research and immunology research.
Catalog No. 50-238-3785