Learn More
ATCC HeLa 229; Cervical Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502383716
Encompass
HeLa 229 cells are human cervical adenocarcinoma cells that were derived from the parent HeLa cell line. They are relatively resistant to polioviruses.
Catalog No. 50-238-3716 Supplier ATCC Supplier No. CCL2.1
HeLa 229 cells are human cervical adenocarcinoma cells that were derived from the parent HeLa cell line. They are relatively resistant to polioviruses.