Learn More
Enzo Life Sciences HeLa, (cell lysate) (200µg)
SDP
Catalog No. 502007333
Encompass_Preferred
Formulation: Liquid. In SDS-PAGE sample buffer. Source: Human. Long Term Storage: -80°C. Handling: Avoid freeze/thaw cycles. Applications: WB .
Catalog No. 50-200-7333 Supplier Enzo Life Sciences Supplier No. ADILYCHL100F
Formulation: Liquid. In SDS-PAGE sample buffer. Source: Human. Long Term Storage: -80°C. Handling: Avoid freeze/thaw cycles. Applications: WB .