Learn More
ATCC HeLa; Cervical Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502382144
Encompass
HeLa is an epithelial cell that was isolated from the cervix of a 31-year-old, Black female with adenocarcinoma. The cell can be used in infectious disease research, sexually transmitted disease research, and bioproduction. The cell line can be used in testing and calibration in ISO 17025-accredited laboratories, to challenge assay performance, validate or compare test methods, and establish sensitivity, linearity, and specificity during assay validation or implementation.
Catalog No. 50-238-2144 Supplier ATCC Supplier No. CRMCCL2
Add to Cart
Add to Cart
HeLa is an epithelial cell that was isolated from the cervix of a 31-year-old, Black female with adenocarcinoma. The cell can be used in infectious disease research, sexually transmitted disease research, and bioproduction. The cell line can be used in testing and calibration in ISO 17025-accredited laboratories, to challenge assay performance, validate or compare test methods, and establish sensitivity, linearity, and specificity during assay validation or implementation.