Learn More
Invivogen HeLa-Difluo hLC3 cells, 3-7 x 10e6 cells
SDP
Supplier:  Invivogen HELDFHLC3/DEL
Encompass
HeLa-Difluo hLC3 reporter cells are designed to monitor the autophagic flux by means of a fluorescent-labeled LC3B protein.
Catalog No. NC1543371