Learn More
Cell Biolabs Inc HeLa/GFP Cell Line
Supplier:  Cell Biolabs Inc AKR213
Encompass_Preferred
Cell Biolabs' HeLa/GFP Cell Line stably expresses GFP and blasticidin-resistant genes. Both GFP and blasticidin-resistant genes were introduced into parental HeLa cervical cancer cells using lentivirus. This cell line otherwise exhibits the same characteristics of the parental HeLa cell line.
Catalog No. 50-109-8817