Learn More
Abcam HeLa Nuclear E x tract, 1MG
Catalog No. 502215680
Encompass_Preferred
Nuclear extract is prepared from HeLa cells, which contains a variety of DNA-binding proteins, transcription factors, and other nuclear proteins.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5680 Supplier Abcam Supplier No. AB2868511MG
Nuclear extract is prepared from HeLa cells, which contains a variety of DNA-binding proteins, transcription factors, and other nuclear proteins.
The product is subject to the following: Abcam Restricted Use Statement