Learn More
Sigma Aldrich Fine Chemicals Biosciences HeLa Nuclear Extract
Catalog No. 502900642
Encompass
Nuclear extract prepared from human HeLa cells using a modified protocol of Dignam et al. which contains a variety of DNA binding proteins and transcription factors.
Catalog No. 50-290-0642 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 12309
Nuclear extract prepared from human HeLa cells using a modified protocol of Dignam et al. which contains a variety of DNA binding proteins and transcription factors.