Learn More
ATCC HeLa S3; Cervical Adenocarcinoma; Human (Homo sapiens)
Catalog No. 50188326FP
Encompass
HeLa S3 cells are a clonal derivative of the parent HeLa cell line (ATCC CCL-2). The HeLa S3 clone has been very useful in the clonal analysis of mammalian cell populations relating to chromosomal variation, cell nutrition, and plaque-forming ability. This line can be adapted to grow in suspension and is a suitable transfection host. The cells are positive for keratin by immunoperoxidase staining. HeLa cells have been reported to contain human papilloma virus 18 (HPV-18) sequences.
Catalog No. 50-188-326FP Supplier ATCC Supplier No. CCL2.2
HeLa S3 cells are a clonal derivative of the parent HeLa cell line (ATCC CCL-2). The HeLa S3 clone has been very useful in the clonal analysis of mammalian cell populations relating to chromosomal variation, cell nutrition, and plaque-forming ability. This line can be adapted to grow in suspension and is a suitable transfection host. The cells are positive for keratin by immunoperoxidase staining. HeLa cells have been reported to contain human papilloma virus 18 (HPV-18) sequences.