Learn More
ATCC HeLaRC32; Cervical Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502381754
Encompass
HeLaRC32 [HeRC32] is an epithelial-like cell that was isolated from the cervix of a Black, 31-year-old patient with adenocarcinoma. This cell line was deposited by P Moullier.
Catalog No. 50-238-1754 Supplier ATCC Supplier No. CRL2972
HeLaRC32 [HeRC32] is an epithelial-like cell that was isolated from the cervix of a Black, 31-year-old patient with adenocarcinoma. This cell line was deposited by P Moullier.