Learn More
Enzo Life Sciences Helenalin (0.5mg). CAS: 6754-13-8
SDP
Supplier:  Enzo Life Sciences ALX350120MC05
Encompass_Preferred
Potent anti-inflammatory and antineoplastic agent. Induces a CD95 death receptor-independent apoptosis even when the mitochondria protecting proteins Bcl-XLor Bcl-2 are overexpressed. Also an inhibitor of NF-κB. Purity: ≥97% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO (10mg/ml) or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-8742