Learn More
Medchemexpress LLC Helenalin acetate 1mg | 10180-86-6 | 304.34 g/mol | 1 MG
SDP
Catalog No. 5000229440
Encompass_Preferred
Helenalin acetate is a sesquiterpene lactone research compound that acts as a natural NF-κB inhibitor and a potent C/EBPβ inhibitor with reported anti-inflammatory and anticancer activity. It is supplied as a solid (white to off-white) at high purity for use in biochemical and cell-based studies.
  • Acts as an NF-κB inhibitor and potent C/EBPβ inhibitor.
  • Reported anti-inflammatory and anticancer activity.
  • High purity (99.55%) suitable for research applications.
  • Supplied in small-mass pack sizes for assay-scale experiments.
  • Store protected from light at -20°C; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-29440 Supplier Medchemexpress LLC Supplier No. HY1145191MG
Helenalin acetate is a sesquiterpene lactone research compound that acts as a natural NF-κB inhibitor and a potent C/EBPβ inhibitor with reported anti-inflammatory and anticancer activity. It is supplied as a solid (white to off-white) at high purity for use in biochemical and cell-based studies.
  • Acts as an NF-κB inhibitor and potent C/EBPβ inhibitor.
  • Reported anti-inflammatory and anticancer activity.
  • High purity (99.55%) suitable for research applications.
  • Supplied in small-mass pack sizes for assay-scale experiments.
  • Store protected from light at -20°C; in solvent: -80°C (6 months), -20°C (1 month).