Learn More
Medchemexpress LLC Helenalin acetate 5mg | 10180-86-6 | 304.34 g·mol⁻¹ | 5 MG
SDP
Catalog No. 5000229441
Encompass_Preferred
Helenalin acetate is a natural sesquiterpene lactone supplied as a research reagent. It inhibits nuclear factor κB (NF-κB) and is a potent C/EBPβ inhibitor, with reported anti-inflammatory and anticancer activities. The compound is provided at high purity for in vitro pathway and cell biology studies.
  • Natural sesquiterpene lactone.
  • Inhibits NF-κB and C/EBPβ transcription factors.
  • Reported anti-inflammatory and anticancer activity.
  • High purity (99.55%).
  • Chemical formula C17H20O5; molecular weight 304.34 g·mol⁻¹.
  • Available in small research pack sizes (e.g., 5 MG).
Catalog No. 50-002-29441 Supplier Medchemexpress LLC Supplier No. HY1145195MG
Helenalin acetate is a natural sesquiterpene lactone supplied as a research reagent. It inhibits nuclear factor κB (NF-κB) and is a potent C/EBPβ inhibitor, with reported anti-inflammatory and anticancer activities. The compound is provided at high purity for in vitro pathway and cell biology studies.
  • Natural sesquiterpene lactone.
  • Inhibits NF-κB and C/EBPβ transcription factors.
  • Reported anti-inflammatory and anticancer activity.
  • High purity (99.55%).
  • Chemical formula C17H20O5; molecular weight 304.34 g·mol⁻¹.
  • Available in small research pack sizes (e.g., 5 MG).