Learn More
Abcam Helenalin, NF-kappaB inhibitor, 500UG
Catalog No. 502704852
Encompass_Preferred
MW 262.3 Da. NF-κB inhibitor. Induces apoptosis. Shows anti-inflammatory and antineoplastic effects. Shows hepatoprotective effects in vivo. Orally active. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4852 Supplier Abcam Supplier No. AB146197500UG
MW 262.3 Da. NF-κB inhibitor. Induces apoptosis. Shows anti-inflammatory and antineoplastic effects. Shows hepatoprotective effects in vivo. Orally active. .
The product is subject to the following: Abcam Restricted Use Statement