Learn More
Grainger Helical Insert 304SS 1/2-13 x 1.5D PK10
Catalog No. 502752482
Encompass_Preferred
The HELI-COIL Helical Insert 304SS 1/2"-13 x 1.5D PK10 is a stainless steel helical coil insert for repairing damaged threads. It has a nominal thread size of 1/2"-13 and an outside diameter of 0.625". The insert is made from 304 stainless steel for corrosion resistance. This is a pack of 10 helical coil inserts that can be installed into a tapped hole to provide strong, durable internal threads. The inserts provide permanent thread repair and reinforcement in metals and plastics. They meet military specification MS15981 and are RoHS compliant.
Catalog No. 50-275-2482 Supplier Grainger Supplier No. 141J04
The HELI-COIL Helical Insert 304SS 1/2"-13 x 1.5D PK10 is a stainless steel helical coil insert for repairing damaged threads. It has a nominal thread size of 1/2"-13 and an outside diameter of 0.625". The insert is made from 304 stainless steel for corrosion resistance. This is a pack of 10 helical coil inserts that can be installed into a tapped hole to provide strong, durable internal threads. The inserts provide permanent thread repair and reinforcement in metals and plastics. They meet military specification MS15981 and are RoHS compliant.