Learn More
Bon Opus Biosciences Recombinant 2019-nCoV Helicase (N-6His-MBP) -50ug
SDP
Supplier:  Bon Opus Biosciences CR8450UG
Encompass_Preferred
Recombinant 2019-nCoV Helicase is produced by our E.coli expression system and the target gene encoding Ala5325-Gln5925 is expressed with a 6His, MBP tag at the N-terminus. Expression system: E. coli. Accession# P0DTD1. Formulation: Supplied as a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# CR84
Catalog No. 50-208-0403