Learn More
TARGETMOL CHEMICALS INC HELICHRYSETIN 5MG
SDP
Catalog No. 502578889
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Helichrysetin has mild anti-HIV-1 PR activity. Helichrysetin has great potentials for development as an anticancer agent it has cytotoxic effect on four selected cancer cell lines A549 MCF-7 Ca Ski and HT-29. purity: 96%
Catalog No. 50-257-8889 Supplier TARGETMOL CHEMICALS INC Supplier No. TN17275MG
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Helichrysetin has mild anti-HIV-1 PR activity. Helichrysetin has great potentials for development as an anticancer agent it has cytotoxic effect on four selected cancer cell lines A549 MCF-7 Ca Ski and HT-29. purity: 96%