Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Helicin | 618-65-5 | 10 MG
SDP

Catalog No. 5000417721
Encompass_Preferred

Helicin is a chemical compound found in Rosaceae, characterized as a moderate syrB inducer. It can be hydrolyzed by the BglY enzyme. This product is intended for research use only.

  • Moderate syrB inducer
  • Hydrolyzable by BglY enzyme
  • Purity: 99.95%
  • Molecular weight: 284.26
  • Chemical formula: C13H16O7
  • Appearance: Solid
  • Color: White to off-white
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-17721 Supplier Medchemexpress LLC Supplier No. HYN706010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Helicin is a chemical compound found in Rosaceae, characterized as a moderate syrB inducer. It can be hydrolyzed by the BglY enzyme. This product is intended for research use only.

  • Moderate syrB inducer
  • Hydrolyzable by BglY enzyme
  • Purity: 99.95%
  • Molecular weight: 284.26
  • Chemical formula: C13H16O7
  • Appearance: Solid
  • Color: White to off-white
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.