Learn More
ATCC Helicobacter hepaticus; Hh-2
Catalog No. 502383500
Encompass
This is a microaerophilic bacterial strain that was isolated from SCID/NCr mouse liver.
Catalog No. 50-238-3500 Supplier ATCC Supplier No. 51448
This is a microaerophilic bacterial strain that was isolated from SCID/NCr mouse liver.